Ethyl 1-methyl-2-oxopyrrolidine-3-carboxylate 95%
Supplier: Apollo Scientific
OR953211-1G OR953211-250MG
APOSOR953211-1GEA 528 EUR
APOSOR953211-1G APOSOR953211-250MG
Ethyl 1-methyl-2-oxopyrrolidine-3-carboxylate 95%
Ethyl 1-methyl-2-oxopyrrolidine-3-carboxylate
Formula: C₈H₁₃NO₃
MW: 171.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501340
CAS Number: 30932-85-5