Ethyl piperidin-2-ylacetate 96%
Supplier: Apollo Scientific
OR953444-1G OR953444-250MG OR953444-5G
APOSOR953444-1GEA 270 EUR
APOSOR953444-1G APOSOR953444-250MG APOSOR953444-5G
Ethyl piperidin-2-ylacetate 96%
Ethyl piperidin-2-ylacetate
Formula: C₉H₁₇NO₂
MW: 171.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752674
CAS Number: 2739-99-3