N-(2-Amino-1,1-dimethylethyl)-n,n-dimethylamine 98%
Supplier: Apollo Scientific
OR953600-1G OR953600-250MG
APOSOR953600-1GEA 364 EUR
APOSOR953600-1G APOSOR953600-250MG
N-(2-Amino-1,1-dimethylethyl)-n,n-dimethylamine 98%
N-(2-Amino-1,1-dimethylethyl)-n,n-dimethylamine
Formula: C₆H₁₆N₂
MW: 116.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05201592
CAS Number: 76936-44-2