2-Chloro-imidazo[1,2-a]pyridine-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR953908-1G OR953908-250MG
APOSOR953908-1GEA 574 EUR
APOSOR953908-1G APOSOR953908-250MG
2-Chloro-imidazo[1,2-a]pyridine-3-carbaldehyde 95%
2-Chloro-imidazo[1,2-a]pyridine-3-carbaldehyde
Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02708012
CAS Number: 131773-23-4