2H-1,3-Benzodioxol-4-ylmethanamine 97%
Supplier: Apollo Scientific
Synonyms: 1,3-Benzodioxole-4-methanamine
OR953947-1G OR953947-250MG
APOSOR953947-1GEA 364 EUR
APOSOR953947-1G APOSOR953947-250MG
2H-1,3-Benzodioxol-4-ylmethanamine 97%
2H-1,3-Benzodioxol-4-ylmethanamine
Formula: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858181
CAS Number: 182634-34-0