1-(4-Methoxyphenyl)ethylamine hydrochloride 98%
Supplier: Apollo Scientific
OR954061-25G OR954061-5G
APOSOR954061-25GEA 464 EUR
APOSOR954061-25G APOSOR954061-5G
1-(4-Methoxyphenyl)ethylamine hydrochloride 98%
1-(4-Methoxyphenyl)ethylamine hydrochloride
Formula: C₉H₁₃NO·ClH
MW: 187.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198855
CAS Number: 90642-63-0