Methyl 4,4-dimethyl-2-oxocyclohexanecarboxylate 95%
Supplier: Apollo Scientific
OR954112-1G OR954112-250MG
APOSOR954112-1GEA 696 EUR
APOSOR954112-1G APOSOR954112-250MG
Methyl 4,4-dimethyl-2-oxocyclohexanecarboxylate 95%
Methyl 4,4-dimethyl-2-oxocyclohexanecarboxylate
Formula: C₁₀H₁₆O₃
MW: 184.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20261465
CAS Number: 32767-46-7