4-Chloro-1,6-dimethyl-1H-pyrazolo[3,4-d]pyrimidine 95%
Supplier: Apollo Scientific
OR954217-1G OR954217-250MG
APOSOR954217-1GEA 432 EUR
APOSOR954217-1G APOSOR954217-250MG
4-Chloro-1,6-dimethyl-1H-pyrazolo[3,4-d]pyrimidine 95%
4-Chloro-1,6-dimethyl-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₇H₇ClN₄
MW: 182.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09863156
CAS Number: 98550-75-5