4-(4-Methylphenyl)-1H-pyrazole 97%
Supplier: Apollo Scientific
OR954547-1G OR954547-250MG
APOSOR954547-1GEA 326 EUR
APOSOR954547-1G APOSOR954547-250MG
4-(4-Methylphenyl)-1H-pyrazole 97%
4-(4-Methylphenyl)-1H-pyrazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03646991
CAS Number: 111016-46-7