4-(4-Methoxyphenyl)-1H-pyrazole 98%
Supplier: Apollo Scientific
OR954562-1G OR954562-250MG
APOSOR954562-1GEA 574 EUR
APOSOR954562-1G APOSOR954562-250MG
4-(4-Methoxyphenyl)-1H-pyrazole 98%
4-(4-Methoxyphenyl)-1H-pyrazole
Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03646977
CAS Number: 111016-45-6