2-Methanesulphonyl-5-methylaniline 95%
Supplier: Apollo Scientific
OR954645-1G OR954645-5G
APOSOR954645-1GEA 219 EUR
APOSOR954645-1G APOSOR954645-5G
2-Methanesulphonyl-5-methylaniline 95%
2-Methanesulphonyl-5-methylaniline
Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11645433
CAS Number: 876494-64-3