2-Formyl-4,5-dimethoxybenzonitrile 95%
Supplier: Apollo Scientific
OR955329-1G OR955329-250MG
APOSOR955329-1GEA 232 EUR
APOSOR955329-1G APOSOR955329-250MG
2-Formyl-4,5-dimethoxybenzonitrile 95%
2-Formyl-4,5-dimethoxybenzonitrile
Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24678322
CAS Number: 1013112-48-5