2,3-Dihydro-1H-indole-2-carboxylic acid methyl ester 95%
Supplier: Apollo Scientific
OR955713-1G OR955713-5G
APOSOR955713-1GEA 255 EUR
APOSOR955713-1G APOSOR955713-5G
2,3-Dihydro-1H-indole-2-carboxylic acid methyl ester 95%
2,3-Dihydro-1H-indole-2-carboxylic acid methyl ester
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865114
CAS Number: 59040-84-5