Methyl 5-methyl-1H-pyrrole-2-carboxylate 96%
Supplier: Apollo Scientific
Warning
OR955843-1G OR955843-5G
APOSOR955843-1GEA 66.7 EUR
APOSOR955843-1G APOSOR955843-5G
Methyl 5-methyl-1H-pyrrole-2-carboxylate 96%
Methyl 5-methyl-1H-pyrrole-2-carboxylate
Formula: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182371
CAS Number: 1194-97-4