4-Chloro-6-(4-methyl-1H-pyrazol-1-yl)pyrimidine 95%
Supplier: Apollo Scientific
OR956043-1G OR956043-250MG
APOSOR956043-1GEA 137 EUR
APOSOR956043-1G APOSOR956043-250MG
4-Chloro-6-(4-methyl-1H-pyrazol-1-yl)pyrimidine 95%
4-Chloro-6-(4-methyl-1H-pyrazol-1-yl)pyrimidine
Formula: C₈H₇ClN₄
MW: 194.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08691760
CAS Number: 1015845-71-2