1,3,5-Trimethyl-1H-pyrazol-4-ol 95%
Supplier: Apollo Scientific
OR957433-100MG OR957433-1G OR957433-250MG
APOSOR957433-100MGEA 69.6 EUR
APOSOR957433-100MG APOSOR957433-1G APOSOR957433-250MG
1,3,5-Trimethyl-1H-pyrazol-4-ol 95%
1,3,5-Trimethyl-1H-pyrazol-4-ol
Formula: C₆H₁₀N₂O
MW: 126.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441530
CAS Number: 89193-22-6