3-(Chloromethyl)phenol 95%
Supplier: Apollo Scientific
Danger
OR957448-1G OR957448-5G
APOSOR957448-1GEA 330 EUR
APOSOR957448-1G APOSOR957448-5G
3-(Chloromethyl)phenol 95%
3-(Chloromethyl)phenol
Formula: C₇H₇ClO
MW: 142.583 g/mol
Boiling Pt: 105…110 °C (0.1 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06658536
CAS Number: 60760-06-7
UN: 3265
ADR: 8,III