4-Bromo-2-(hydroxymethyl)benzoic acid 95+%
Supplier: Apollo Scientific
Warning
OR957980-100MG OR957980-250MG
APOSOR957980-100MGEA 30.6 EUR
APOSOR957980-100MG APOSOR957980-250MG
4-Bromo-2-(hydroxymethyl)benzoic acid 95+%
4-Bromo-2-(hydroxymethyl)benzoic acid
Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977384
CAS Number: 670256-21-0