Ethyl 4-Amino-3-bromo-5-chlorobenzoate 97%
Supplier: Apollo Scientific
Warning
OR958889-1G OR958889-250MG OR958889-5G
APOSOR958889-1GEA 33.5 EUR
APOSOR958889-1G APOSOR958889-250MG APOSOR958889-5G
Ethyl 4-Amino-3-bromo-5-chlorobenzoate 97%
Ethyl 4-Amino-3-bromo-5-chlorobenzoate
Formula: C₉H₉BrClNO₂
MW: 278.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03407826
CAS Number: 874779-56-3