1-Benzyl-1H-pyrazol-4-amine hydrochloride
Supplier: Apollo Scientific
Warning
OR959336-1G OR959336-5G
APOSOR959336-1GEA 21.9 EUR
APOSOR959336-1G APOSOR959336-5G
1-Benzyl-1H-pyrazol-4-amine hydrochloride
1-Benzyl-1H-pyrazol-4-amine hydrochloride
Formula: C₁₀H₁₁N₃·ClH
MW: 209.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18425906
CAS Number: 1264097-17-7