Methyl 2-methoxy-5-methylpyridine-3-carboxylate 95%
Supplier: Apollo Scientific
OR959608-1G OR959608-250MG
APOSOR959608-1GEA 632 EUR
APOSOR959608-1G APOSOR959608-250MG
Methyl 2-methoxy-5-methylpyridine-3-carboxylate 95%
Methyl 2-methoxy-5-methylpyridine-3-carboxylate
Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611638
CAS Number: 351410-41-8