2,4-Dimethoxypyrimidine-5-carbonitrile 97%
Supplier: Apollo Scientific
OR959612-100MG OR959612-1G OR959612-250MG
APOSOR959612-100MGEA 203 EUR
APOSOR959612-100MG APOSOR959612-1G APOSOR959612-250MG
2,4-Dimethoxypyrimidine-5-carbonitrile 97%
2,4-Dimethoxypyrimidine-5-carbonitrile
Formula: C₇H₇N₃O₂
MW: 165.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24448829
CAS Number: 14445-57-9