Isoxazolo[5,4-c]pyridin-3-amine 98%
Supplier: Apollo Scientific
OR959629-1G OR959629-250MG
APOSOR959629-1GEA 178 EUR
APOSOR959629-1G APOSOR959629-250MG
Isoxazolo[5,4-c]pyridin-3-amine 98%
Isoxazolo[5,4-c]pyridin-3-amine
Formula: C₆H₅N₃O
MW: 135.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193315
CAS Number: 114080-94-3