3-(Methoxycarbonyl)-4-methylbenzoic acid 95%
Supplier: Apollo Scientific
OR959638-1G OR959638-250MG
APOSOR959638-1GEA 203 EUR
APOSOR959638-1G APOSOR959638-250MG
3-(Methoxycarbonyl)-4-methylbenzoic acid 95%
3-(Methoxycarbonyl)-4-methylbenzoic acid
Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03265441
CAS Number: 167300-06-3