2,3-Dichloropyrido[1,2-a]pyrimidin-4-one 96%
Supplier: Apollo Scientific
Warning
OR959640-1G OR959640-5G
APOSOR959640-1GEA 377 EUR
APOSOR959640-1G APOSOR959640-5G
2,3-Dichloropyrido[1,2-a]pyrimidin-4-one 96%
2,3-Dichloropyrido[1,2-a]pyrimidin-4-one
Formula: C₈H₄Cl₂N₂O
MW: 215.036 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD25953923
CAS Number: 117890-82-1