1-(5-hydroxypyridin-2-yl)ethanone 95%
Supplier: Apollo Scientific
Synonyms: 1-(5-Hydroxypyridin-2-yl)ethanone
OR959785-1G
APOSOR959785-1GEA 128 EUR
APOSOR959785-1G
1-(5-hydroxypyridin-2-yl)ethanone 95%
1-(5-hydroxypyridin-2-yl)ethanone
Formula: C₇H₇NO₂
MW: 137.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697710
CAS Number: 67310-56-9