1-Isobutylpyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
Synonyms:
1-Isobutyl-1H-pyrazole-4-carbaldehyde
OR960037-1G
OR960037-5G
APOSOR960037-1GEA
72.5
EUR
APOSOR960037-1G
APOSOR960037-5G
1-Isobutylpyrazole-4-carbaldehyde 97%
1-Isobutylpyrazole-4-carbaldehyde
Formula:
C₈H₁₂N₂O MW: 152.194 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08559037 CAS Number: 1006333-32-9 |