4-(Propane-1-sulphonyl)aniline 98%
Supplier: Apollo Scientific
OR960090-1G OR960090-5G
APOSOR960090-1GEA 219 EUR
APOSOR960090-1G APOSOR960090-5G
4-(Propane-1-sulphonyl)aniline 98%
4-(Propane-1-sulphonyl)aniline
Formula: C₉H₁₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09936494
CAS Number: 86810-78-8