1-Cyclobutylethan-1-amine hydrochloride
Supplier: Apollo Scientific
Synonyms: 1-Cyclobutylethanamine hydrochloride
OR960119-1G OR960119-250MG OR960119-500MG
APOSOR960119-1GEA 1110 EUR
APOSOR960119-1G APOSOR960119-250MG APOSOR960119-500MG
1-Cyclobutylethan-1-amine hydrochloride
1-Cyclobutylethan-1-amine hydrochloride
Formula: C₆H₁₃N·ClH
MW: 135.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690555
CAS Number: 904733-73-9