(3E)-4-(2-Methylpyrazol-3-yl)but-3-en-2-one 98%
Supplier: Apollo Scientific
OR960120-1G OR960120-250MG
APOSOR960120-1GEA 632 EUR
APOSOR960120-1G APOSOR960120-250MG
(3E)-4-(2-Methylpyrazol-3-yl)but-3-en-2-one 98%
(3E)-4-(2-Methylpyrazol-3-yl)but-3-en-2-one
Formula: C₈H₁₀N₂O
MW: 150.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383784
CAS Number: 1393444-14-8