(2-Methyl-2H-pyrazol-3-yl)acrylic acid 98%
Supplier: Apollo Scientific
OR960121-10G OR960121-1G OR960121-5G
APOSOR960121-10GEA 502 EUR
APOSOR960121-10G APOSOR960121-1G APOSOR960121-5G
(2-Methyl-2H-pyrazol-3-yl)acrylic acid 98%
(2-Methyl-2H-pyrazol-3-yl)acrylic acid
Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08559038
CAS Number: 1006959-18-7