7-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Supplier: Apollo Scientific

OR960205-1G OR960205-5G
APOSOR960205-1GEA 219 EUR
APOSOR960205-1G APOSOR960205-5G
7-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine 98%
7-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine
Formula: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD20687056
CAS Number: 1173183-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR