4,7-Dichloro-1H-indole 96%
Supplier: Apollo Scientific
OR960257-1G OR960257-250MG
APOSOR960257-1GEA 531 EUR
APOSOR960257-1G APOSOR960257-250MG
4,7-Dichloro-1H-indole 96%
4,7-Dichloro-1H-indole
Formula: C₈H₅Cl₂N
MW: 186.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749917
CAS Number: 96129-73-6