3-Methyl-5-(4-methylphenyl)-1,2,4-oxadiazole 98%

Supplier: Apollo Scientific

OR960285-1G OR960285-250MG
APOSOR960285-1GEA 632 EUR
APOSOR960285-1G APOSOR960285-250MG
3-Methyl-5-(4-methylphenyl)-1,2,4-oxadiazole 98%
3-Methyl-5-(4-methylphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13961847
CAS Number: 122503-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR