(Z)-3-Bromoacrylic acid 98%
Supplier: Apollo Scientific
OR960389-1G OR960389-250MG
APOSOR960389-1GEA 328 EUR
APOSOR960389-1G APOSOR960389-250MG
(Z)-3-Bromoacrylic acid 98%
(Z)-3-Bromoacrylic acid
Formula: C₃H₃BrO₂
MW: 150.959 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075363
CAS Number: 1609-92-3