5-Chloro-2,3-dihydro-1H-inden-1-ol 95%

Supplier: Apollo Scientific

OR960393-1G OR960393-250MG
APOSOR960393-1GEA 79.9 EUR
APOSOR960393-1G APOSOR960393-250MG
5-Chloro-2,3-dihydro-1H-inden-1-ol 95%
5-Chloro-2,3-dihydro-1H-inden-1-ol
Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11219500
CAS Number: 33781-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR