2-Naphthalen-2-yl-ethylamine 96%
Supplier: Apollo Scientific
OR961210-1G OR961210-250MG OR961210-5G
APOSOR961210-1GEA 290 EUR
APOSOR961210-1G APOSOR961210-250MG APOSOR961210-5G
2-Naphthalen-2-yl-ethylamine 96%
2-Naphthalen-2-yl-ethylamine
Formula: C₁₂H₁₃N
MW: 171.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01529868
CAS Number: 2017-68-7