10-Chlorocamphor 95%
Supplier: Apollo Scientific
OR961552-1G OR961552-5G
APOSOR961552-1GEA 219 EUR
APOSOR961552-1G APOSOR961552-5G
10-Chlorocamphor 95%
10-Chlorocamphor
Formula: C₁₀H₁₅ClO
MW: 186.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20261478
CAS Number: 51057-38-6