2-(1,4-Dioxa-spiro[4.5]dec-8-yl)-ethylamine 95%
Supplier: Apollo Scientific
OR961896-1G OR961896-250MG
APOSOR961896-1GEA 864 EUR
APOSOR961896-1G APOSOR961896-250MG
2-(1,4-Dioxa-spiro[4.5]dec-8-yl)-ethylamine 95%
2-(1,4-Dioxa-spiro[4.5]dec-8-yl)-ethylamine
Formula: C₁₀H₁₉NO₂
MW: 185.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114534
CAS Number: 124499-34-9