1-Hydroxypiperidine 95%
Supplier: Apollo Scientific
Synonyms: Piperidin-1-ol
OR961933-1G OR961933-5G
APOSOR961933-1GEA 85.7 EUR
APOSOR961933-1G APOSOR961933-5G
1-Hydroxypiperidine 95%
1-Hydroxypiperidine
Formula: C₅H₁₁NO
MW: 101.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038048
CAS Number: 4801-58-5