3(5)-tert-Butyl-4-nitro-1H-pyrazole 98%
Supplier: Apollo Scientific
OR962475-1G OR962475-5G
APOSOR962475-1GEA 248 EUR
APOSOR962475-1G APOSOR962475-5G
3(5)-tert-Butyl-4-nitro-1H-pyrazole 98%
3(5)-tert-Butyl-4-nitro-1H-pyrazole
Formula: C₇H₁₁N₃O₂
MW: 169.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127341
CAS Number: 38858-95-6