6-(2-Thienyl)-2-pyridinecarboxaldehyde 95%

Supplier: Apollo Scientific

APOSOR963394-1GEA 256 EUR
APOSOR963394-1G
6-(2-Thienyl)-2-pyridinecarboxaldehyde 95%
6-(2-Thienyl)-2-pyridinecarboxaldehyde
Formula: C₁₀H₇NOS
MW: 189,234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410007
CAS Number: 208111-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR