2,3,6,7-Tetramethylpyrazolo[1,5-a]pyrimidin-5-amine 95%
Supplier: Apollo Scientific
OR963525-1G OR963525-5G
APOSOR963525-1GEA 161 EUR
APOSOR963525-1G APOSOR963525-5G
2,3,6,7-Tetramethylpyrazolo[1,5-a]pyrimidin-5-amine 95%
2,3,6,7-Tetramethylpyrazolo[1,5-a]pyrimidin-5-amine
Formula: C₁₀H₁₄N₄
MW: 190.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19382365
CAS Number: 1311254-54-2