2-Chloro-4-methoxy-3-nitropyridine 98%
Supplier: Apollo Scientific
OR963856-10G OR963856-1G OR963856-5G
APOSOR963856-10GEA 217 EUR
APOSOR963856-10G APOSOR963856-1G APOSOR963856-5G
2-Chloro-4-methoxy-3-nitropyridine 98%
2-Chloro-4-methoxy-3-nitropyridine
Formula: C₆H₅ClN₂O₃
MW: 188.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00955756
CAS Number: 6980-09-2