Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate 95%
Supplier: Apollo Scientific
OR964339-1G OR964339-250MG
APOSOR964339-1GEA 265 EUR
APOSOR964339-1G APOSOR964339-250MG
Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 1-(tert-butyl)-1H-pyrazole-4-carboxylate
Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17206126
CAS Number: 139308-53-5