3,3-Dimethyl-tetrahydro-pyran-2-one 97%
Supplier: Apollo Scientific
Synonyms: 3,3-Dimethyltetrahydro-2H-pyran-2-one
OR964411-1G OR964411-250MG
APOSOR964411-1GEA 478 EUR
APOSOR964411-1G APOSOR964411-250MG
3,3-Dimethyl-tetrahydro-pyran-2-one 97%
3,3-Dimethyl-tetrahydro-pyran-2-one
Formula: C₇H₁₂O₂
MW: 128.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369942
CAS Number: 4830-05-1