3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%
Supplier: Apollo Scientific
Synonyms: 3-(2-(3-Ethoxy-3-oxopropoxy)ethoxy)propanoic acid, Acid-PEG2-ethyl propionate
OR964496-1G OR964496-250MG
APOSOR964496-1GEA 348 EUR
APOSOR964496-1G APOSOR964496-250MG
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid
Formula: C₁₀H₁₈O₆
MW: 234.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115349
CAS Number: 886362-90-9