4-Isopropyloxazolidine-2,5-dione 95%
Supplier: Apollo Scientific
OR964676-1G OR964676-5G
APOSOR964676-1GEA 177 EUR
APOSOR964676-1G APOSOR964676-5G
4-Isopropyloxazolidine-2,5-dione 95%
4-Isopropyloxazolidine-2,5-dione
Formula: C₆H₉NO₃
MW: 143.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00196817
CAS Number: 2816-12-8