2-Chloro-1-(2,4,6-trimethyl-phenyl)-ethanone 95%
Supplier: Apollo Scientific
OR964768-1G OR964768-5G
APOSOR964768-1GEA 502 EUR
APOSOR964768-1G APOSOR964768-5G
2-Chloro-1-(2,4,6-trimethyl-phenyl)-ethanone 95%
2-Chloro-1-(2,4,6-trimethyl-phenyl)-ethanone
Formula: C₁₁H₁₃ClO
MW: 196.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00507868
CAS Number: 50690-12-5