4-[(Pyridin-2-yl)oxy]benzaldehyde 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Formylphenoxy)pyridine
OR9651-1G OR9651-5G
APOSOR9651-1GEA 63.8 EUR
APOSOR9651-1G APOSOR9651-5G
4-[(Pyridin-2-yl)oxy]benzaldehyde 97+%
4-[(Pyridin-2-yl)oxy]benzaldehyde
Formula: C₁₂H₉NO₂
MW: 199.205 g/mol
Melting Pt: 45…46 °C
Storage Temperature: Ambient
MDL Number: MFCD08435899
CAS Number: 194017-69-1